Aprons, PE, Hopen®
Leverancier: Medicom
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
50.601 50.101 50.201
HOPE50.601EA 12.8 EUR
HOPE50.601 HOPE50.101 HOPE50.201
Aprons, PE, Hopen®
Wegwerpkleding
These disposable aprons made of embossed PE protect body and clothing from dust, dirt and liquids.
  • Highly resistant ties around neck and waist
  • Wall-mountable polybags allow a 'one by one' dispensing which prevents contamination
  • Available in two different sizes
Ideal for use in laboratories, medical environments and for food processing.
Order Now
Specificaties
Environmentally Preferable
Geslacht
Kledingstuk toepassing Food