Bouffant caps, Hopen®
Leverancier: Medicom
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
70.112 70.113 70.111
HOPE70.112EA 30.4 EUR
HOPE70.112 HOPE70.113 113-8613
Bouffant caps, Hopen®
Wegwerpkleding
These disposable bouffant caps made of PP are ideal for use in hygiene and food industries.
  • Cover the hair as required by HACCP standards
  • Double elasticated for comfort and perfect fit
  • Available in different colours
  • Packed in convenient dispenser box which can also hold other items
Weight: 12 g/m²