Metering valves, Accuflo™
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MV4-4F MV4-4N MV4-4P MV4-4Q MV4-6F MV4-6Q
ENTEMV4-4FEA 0 EUR
ENTEMV4-4F ENTEMV4-4N ENTEMV4-4P ENTEMV4-4Q ENTEMV4-6F ENTEMV4-6Q
Metering valves, Accuflo™
Ventielen Overdrukventielen
These components are constructed of high purity PFA and PTFE materials for wetted parts and PVDF, PEEK™ and PTFE u-cup with Viton® energizer for non wetted parts.
  • Multiple turn with manual adjustment
  • Standard panel mounting configuration
  • Ultrapure and corrosion-resistant performance
Maximum continuous use pressure: 689 kPa at ambient temperature.
Maximum continuous use temperature: 149 °C at 172 kPa.