Tubings, PFA, high purity, PureBond® 4200
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PA500 PA2000MTR3 PA250MTR3 PA1000 PA1000MTR3 PA500MTR3_U PA750MTR3 PA2000X PA250X PA250 PA750X PA500X PA750
ENTEPA500EA 0 EUR
ENTEPA500 ENTEPA2000MTR3 ENTEPA250MTR3 ENTEPA1000 ENTEPA1000MTR3 ENTEPA500MTR3_U ENTEPA750MTR3 ENTEPA2000X ENTEPA250X ENTEPA250 ENTEPA750X ENTEPA500X ENTEPA750
Tubings, PFA, high purity, PureBond® 4200
Slangen Transfer Tubing
Made of PFA, DuPont 440.
Permanent serialized laser marking is a nondestructive and noncontaminating method to identify tube material and Ø. It also allows for 100% traceability to a specific material lot, production date and production inspection record.