Viabank™ Bacterial Storage Beads
Leverancier: MEDICAL WIRE EQUIPMENT
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
MWVIB
MWVIG
MWVIY
MWVIR
MWVIM
MEDWMWVIBEA
305
EUR
MEDWMWVIB
MEDWMWVIG
MEDWMWVIY
MEDWMWVIR
MEDWMWVIM
Viabank™ Bacterial Storage Beads
Testsystemen voor microbiologie
Micro-organisme testen
Viabank™ bacterial storage beads for long term storage of microorganisms at low temperatures.
- Twenty glass beads per vial
- Coloured caps match colour of beads
- Available in multi coloured packs of red, yellow, green and blue or singular packs of each colour
- Individual beads can be removed while frozen at any time for establishing fresh cultures