Scalpels with metal handle, GRAFIX SMALL / LARGE
Leverancier: MARTOR ARGENTAX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
23111.54 23112.54
MARO23111.54EA 533 EUR
MARO23111.54 MARO23112.54
Scalpels with metal handle, GRAFIX SMALL / LARGE
Scalpels Scalpels
These durable scalpels entirely made of chrome plated steel are ideal for work on graphic paper, film sheets and textiles. Supplied with either a small or a large blade with rounded off cutting edge.
  • Full metal housing: Very abrasion resistant
  • Handle can be used with different blades
  • Blade can easily be exchanged (without tools)
  • Lightweight, ergonomic metal handle with grooves for a better grip
  • Can be used by left and right handers
Leveringsinformatie: GRAFIX SMALL is supplied with blade No. 13, GRAFIX LARGE is supplied with blade No. 21.