Magnetic strips, B-859
Leverancier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
058301 225172 225181
BRDY058301EA 30.8 EUR
BRDY058301 BRDY225172 BRDY225181
Magnetic strips, B-859
Etiketten
Helps to mark and identify racking and shelving quickly and easily with this flexible magnetic strip.
  • Relocation of the marker when required
  • Keeps the face of the rack clean in appearance
Use the wipe-off pen to write on these strips.