Cable ties, T, PA66
Leverancier: HELLERMANN TYTON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
111-01919 111-01626 111-02370
129-4850EA 31.3 EUR
129-4850 129-4851 HELT111-02370
Cable ties, T, PA66
Spanbanden
These T-series cable ties in polyamide 6.6 are used for routing, bundling and securing of cables, pipes and hoses.
  • High tensile strength with very low insertion force
  • Serrated inside for a strong hold onto bundles
  • Ergonomic bent tail offers quick and simple installation
  • Easy application either manually or with an application tool
The cable ties can be used for nearly all kind of applications in the solar energy, wind energy and rail transportation sector, electronics, defence, aerospace and shipbuilding industry.