Jugs, with handle
Leverancier: EP SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
115-4000 315-4000 315-2000
EPSC115-4000EA 90 EUR
EPSC115-4000 EPSC315-4000 EPSC315-2000
Jugs, with handle
Flessen Voorraadflessen
HDPE, natural, with white LDPE foam-lined PP screw closure.
  • Good chemical resistance
  • Good temperature resistance
The narrow mouth and sturdy handle make carrying and pouring higher volume samples easy. These are recommended for collecting water samples to be analyzed for inorganic trace analytes. Unprocessed or certified available.
Meet or exceed EPA performance-based standards for metals, cyanide and fluoride.