Spectral lamps, OSRAM
Leverancier: DIDALAB
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
POD068380 POD068360 POD068400 POD068420 POD068440 POD068450 POD068480
DIDAPOD068380EA 783 EUR
DIDAPOD068380 DIDAPOD068360 DIDAPOD068400 DIDAPOD068420 DIDAPOD068440 DIDAPOD068450 DIDAPOD068480
Spectral lamps, OSRAM
Lampen Spectrumlampen
Nine pin socket spectral lamps with dimensions (Hר) 107×21 mm, operated with 1 Amp fixed current.