Aluminium crimp caps
Leverancier: THERMO NATIONAL SCIENTIFIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C4008-1A C4008-2A C4008-4A
NSCAC4008-1AEA 50.6 EUR
NSCAC4008-1A NSCAC4008-2A NSCAC4008-4A
Aluminium crimp caps
Sluitingen Sluitingen voor chromatografievials
Pre-assembled caps and septa are convenient and minimise contamination from handling.
  • Aluminium crimp caps with prefitted septa
  • Provide a secure leak-resistant seal