Safety storage cabinets, type 30, Q-CLASSIC-30
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
31003069 31003068 31002060 31003066 31007061 31003061 31003060 31002070 31002069 31002068 31002067 31002066 31002061 31003067 31008066 31009069 31009068 31009067 31009066 31009061 31009060 31008070 31008069 31003070 31008067 31007060 31008061 31008060 31007070 31007069 31007068 31007067 31007066 31009070 31008068
135-2474EA 2480 EUR
135-2474 135-2473 135-1925 135-2471 135-2477 135-1922 135-1924 135-2470 135-2469 135-2468 135-2467 135-2466 135-1926 135-2472 135-2483 135-2493 135-2492 135-2491 135-2490 135-2489 135-2488 135-2487 135-2486 135-2475 135-2484 135-2476 135-1920 135-1919 135-2482 135-2481 135-2480 135-2479 135-2478 135-2494 135-2485
Safety storage cabinets, type 30, Q-CLASSIC-30
Meubilair Opslagkasten
Fireproof safety cabinets, classified type 30 (fire resistant up to 30 minutes), safe, durable, robust and convenient with transport base and bicoloured cabinet design.
  • Robust construction and longevity – triple hinged door, safety elements assembled outside the storage compartment for increased protection against corrosion, scratch and impact resistant surface, easy to clean
  • Easy handling with comfort – doors stop in any position, self-closing in the event of fire
  • No unauthorised use – doors lockable with cylinder locking (integration in a locking system possible) and locking state indicator (red/green)
  • Easy transport – integrated transport base with removable, optional base cover
  • Easy alignment – adjusting aids to compensate for uneven floor
  • Quickly put into operation – integrated exhaust outlet ready for connection (NW 75 mm) to a technical exhaust system
The two-colour look with a choice of 7 door colours, at no extra cost, offers maximum individuality and a modern appearance.
Certificaten: Furnace tested (type test) in accordance with EN 14470-1
GS approval, CE conformity
EN 14727 conformity (laboratory furniture)
Bestelinformatie: Seven door colours are available. Supplied without interior equipment packages. Interior equipment packages need to be ordered separately. Please check the website or contact your local Avantor office for further information.
Leveringsinformatie: Delivery prices available upon request.