Tubing connectors, mini
Leverancier: PIEPER FILTER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PP-6MS16 PP-6MS24 PP-6MS32 PP-6MS40 PP-6MS48
229-4111EA 75 EUR
229-4111 229-4112 229-4113 229-4114 229-4115
Tubing connectors, mini
Fittingen en connectoren
PP, translucent.
  • Thread M 6 with tubing olive