Safety storage cabinets for chemicals, CS-Classic
Leverancier: asecos
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
30495040 30495046 30495047 30495048 30495049 30495050
MSGA30495040EA 1110 EUR
MSGA30495040 MSGA30495046 MSGA30495047 MSGA30495048 MSGA30495049 MSGA30495050
Safety storage cabinets for chemicals, CS-Classic
Meubilair Opslagkasten
Storage of non-flammable chemicals and toxic substances in workroom.
  • Robust construction and longevity – body made of sheet steel with highgrade structured paint finish
  • No unauthorised use – doors lockable with cylinder locking (integration in a locking system possible)
  • Easy transport – integrated transport base with removable, optional base cover
  • Easy alignment – adjusting aids to compensate for uneven floor
  • Five more door colours available upon request at no extra cost
Certificaten: TÜV approved.
Bestelinformatie: Interior equipment packages needMore Product Information