Cooling water tubing
Leverancier: LAUDA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RKJ033 RKJ031 RKJ103 RKJ104 RKJ105 RKJ032
LAUDRKJ033EA 44.1 EUR
LAUDRKJ033 LAUDRKJ031 LAUDRKJ103 LAUDRKJ104 LAUDRKJ105 LAUDRKJ032
Cooling water tubing
Slangen Transfer Tubing
EPDM tubing, reinforced.
Special polymer tubings for high pressure. Not suitable for Ultra 350 and mineral oils.