Heat sealing foil, PeelASeal Foil™
Leverancier: iST Scientific Ltd
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IST-104-078SS IST-104-115TR IST-104-115SR IST-104-115LR IST-104-078TR IST-104-078SR IST-104-078TS IST-104-078LS IST-104-078LR
731-0552EA 111 EUR
731-0552 731-0554 731-0550 731-0549 731-0553 731-0548 731-0555 731-0551 731-0547
Heat sealing foil, PeelASeal Foil™
Afdichtingen Afdichtingen van microtiterplaten
Peelable foil laminate for polyproylene plates. Suitable for PCR, low temperature, short term compound storage, short term room temperature compound storage (less than 5 days).
  • Peelable (from −80 °C)
  • Non pierceable
  • Moderate solvent resistance with good liquid barrier
  • Seal integrity range: −80 to 110 °C
Order Now
Specificaties
Environmentally Preferable