Heat sealing foil, PeelASeal DMSO Foil™
Leverancier: iST Scientific Ltd
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IST-105-078SS IST-105-078SR IST-105-078LS IST-105-078LR IST-105-078TR IST-105-115TR IST-105-078TS IST-105-115SR IST-105-115LR
731-0561EA 118 EUR
731-0561 731-0557 731-0560 731-0556 731-0562 731-0563 731-0564 731-0559 731-0558
Heat sealing foil, PeelASeal DMSO Foil™
Afdichtingen Afdichtingen van microtiterplaten
Peelable DMSO resistant foil for polypropylene plates. Suitable for low temperature and ambient temperature storage with DMSO and other solvents.
  • Peelable
  • Non pierceable
  • High solvent resistance with good liquid barrier
  • Seal integrity range: −80 to 80 °C