Heat sealing foil, PierceASeal Foil PS™
Leverancier: iST Scientific Ltd
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IST-107-078SR IST-107-078LR IST-107-078SS IST-107-078TR IST-107-078TS IST-107-115LR IST-107-115SR IST-107-115TR IST-107-078LS IST-107-200LR
731-0575EA 836 EUR
731-0575 731-0574 731-0579 731-0580 731-0582 731-0576 731-0577 731-0581 731-0578 ISTSIST-107-200LR
Heat sealing foil, PierceASeal Foil PS™
Afdichtingen Afdichtingen van microtiterplaten
Pierceble and peelable, high grade foil for polystyrene plates with good solvent resistance including DMSO. Suitable for PCR low temperature compound storage, short term room temperature compound storage.
  • Peelable to polystyrene plates only
  • Multiple sealing and resealing permissible
  • Easy non sealing surface identification
  • Seal integrity range: −20 to 110 °C