Heat sealing foil, Gas PermASeal™
Leverancier: iST Scientific Ltd
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IST-110-115SR IST-110-115TR IST-110-078TS IST-110-078TR IST-110-115LR IST-110-078SS IST-110-078SR IST-110-078LS IST-110-078LR
731-0604EA 858 EUR
731-0604 731-0608 731-0609 731-0607 731-0603 731-0606 731-0602 731-0605 731-0601
Heat sealing foil, Gas PermASeal™
Afdichtingen Afdichtingen van microtiterplaten
Non woven gas permeable and peelable seal for PP and PS plates. Suitable for short term incubation, agriculture and seed storage, insect storage, cell culture.
  • Pierceable and peelable
  • Barriers to solid components
  • Porous and opaque
  • Seal integrity range: −20 to +80 °C
  • Not certified free from nucleases and DNA