Bunsen burners, LabFlame
Leverancier: BioTool
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
LF8.200.000 LF6.004.000
BTOLLF8.200.000EA 0 EUR
BTOLLF8.200.000 BTOLLF6.004.000
Bunsen burners, LabFlame
Verwarmers Gasbranders
Stainless steel Bunsen burners, with removable burner head. The optimised air-flow system allows a uniform and constant flame even in safety cabinets with horizontal or vertical air flow. All controls for gas and air supply as well as the multifunctional control for the modes of operation are conveniently located on the easy to read display.
  • Can be connected to any kind of gas source
  • Optimised for use in microbiology laboratories
  • Extremely firm and consistent flame produced with only a few flicks of the wrist