Adhesive sealing film for microplates, QuickSeal DMSO X Film™
Leverancier: iST Scientific Ltd
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
IST-126-080TS IST-126-080LS IST-126-080SS
731-0631EA 20.4 EUR
731-0631 731-0629 731-0630
Adhesive sealing film for microplates, QuickSeal DMSO X Film™
Afdichtingen Afdichtingen van microtiterplaten
Transparent DMSO resistant film with piercing X. Suitable for sample access and retrieval for 96 well plates for use with autosamplers and sequencers.
  • Peelable with crosscuts over the wells
  • Automatically cleans tips on extraction
  • Multiple sealing and resealing permissible
  • Seal integrity range: −40 to +80 °C