Urine test strips, urinalysis
Leverancier: SURESCREEN
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
RS10/100
RS11/50
RS2/50
RS7/25
RS8/100
RS8/25
RS8/50
SURERS10/100EA
16.6
EUR
SURERS10/100
SURERS11/50
SURERS2/50
SURERS7/25
SURERS8/100
SURERS8/25
SURERS8/50
Urine test strips, urinalysis
Diagnostische systemen
Clinical Reagents and Kits
To determine health function from a urine sample. Parameters can include:
- Ascorbic acid
- Glucose
- Bilirubin
- Ketones
- Specific gravity
- Blood
- pH
- Protein
- Urobilinogen
- Nitrates
- Leukocytes
The test strip features various colour change pads that indicate correct concentrations of various markers in a urine sample. Test strip is submerged in a urine sample for a few seconds before being compared with the results chart to measure any change in colour.