Test charts
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SIMEVF2319EA 123 EUR
SIMEVF2319 SIMEVF2316 SIMEVF2317 SIMEVF2325 SIMEVF2347 SIMEVF3007 SIMEVF3008 SIMEVF2345 SIMEVF3005 SIMEVF2323 SIMEVF2346 SIMEVF3006 SIMEVF2354 SIMEVF2343 SIMEVF3003 SIMEVF2321 SIMEVF3004 SIMEVF2344 SIMEVF3001 SIMEVF3002 SIMEVF3000
Test charts
Kaarten
Test charts are used for testing physical properties of coating, lacquers, and inks.
  • Dimensions from A6 up to A4
  • Film laminated for an excellent solvent, chemical resistance, and film spread
  • Section for the date, time, and test number
Suitable for determining hiding power, opacity, and spreading rate in coating laboratories and paint production.
Bestelinformatie: A certificate is available for charts with an optical brightener.