Cleanroom handschoenen, nitril, AdvanTech 519
Leverancier: MAPA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
34519017 34519018 34519309 34519010 34519019
112-4905EA 401 EUR
112-4905 112-4904 112-4903 112-4907 MAPA34519019
Cleanroom handschoenen, nitril, AdvanTech 519
Handschoenen
These disposable gloves combine chemical protection with excellent mechanical resistance. Recommended for people sensitive to the proteins in natural latex.
  • Non sterile, chlorinated, textured finish
  • Manufacturing process guarantees a very low level of contaminants (particles, non-volatile residue, extractables)
  • Optimal dexterity without compromising chemical resistance
  • Better resistance than natural latex gloves against organic solvents
Suitable for use in Class 100, ISO 5 or M3.5 cleanrooms.
Certificaten: EN 388:2016 (2001X), EN ISO 374-1 Type B (JOT), EN ISO 374-5; Cat. III.