Safety Spectacles, Silium+
Leverancier: Bollé Safety
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SILPPSI SILPPSF
111-1625EA 13.33 EUR
111-1625 111-1627
Safety Spectacles, Silium+
Brillen
Inspired from SILIUM, SILIUM + are comfortable and ultra lightweight PC safety glasses. SILIUM + is an excellent choice, combining style, ergonomics and performances for extended use.
  • Adjustable bridge
  • Non-slip bridge
  • Straight, highly flexible ergonomic temples
  • PLATINUM coating
Weight: 21 g
Certificaten: EN 166 FT1, EN 170, EN 172 (smoke lens).