Precast gels, Reliant™
Leverancier: LONZA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
54925 54803 54922 54903 54927 54929 54801 54813 54820 54905 54907 54928 54939 54944 54948
733-1567EA 308 EUR
733-1567 733-1564 733-1566 733-1565 733-1568 733-1569 733-1570 733-1571 733-1572 733-1576 733-1577 733-1579 733-1581 733-1582 733-1585
Precast gels, Reliant™
Gel voor elektroforese Voorgegoten gels
Reliant™ precast agarose minigels are designed for rapid and reproducible resolution of DNA fragments from 8 bp to 10 kb. Each precast agarose gel is precision cast using our SeaKem® Gold or NuSieve™ 3:1 agarose and are available in a variety of well formats and agarose concentrations.
  • Gels are cast from either SeaKem® or NuSieve® agarose and are certified DNase-free
  • Gels specifically for RNA separation are guaranteed RNase-free
  • Compatible with most minigel chambers
Reliant™ precast agarose minigels are available either pre-stained with ethidiumMore Product Information