Fluoroline® Ultrapure PFA Tubings
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AT500-062 AT250-047 AT125-030_U AT750-062 AT125-030
222-0007EA 0 EUR
222-0007 222-0005 222-0004 ENTEAT750-062 ENTEAT125-030
Fluoroline® Ultrapure PFA Tubings
Slangen Transfer Tubing
Made of PFA, DuPont 440.
  • Compliant to SEMI Standard F57-0301, the specification for polymer components used in ultrapure water and liquid chemical distribution systems
  • Made of 100% virgin high purity material to provide the lowest extractable ionic and organic levels in PFA materials
  • Tubing is produced with specially designed extruders and extrusion processes
  • Tight and consistent tolerances, along with lot-to-lot consistency, are maintained using continuous on-line measurements of the FluoroLine® tubing ext. Ø. and wall thickness
Permanent serialisedMore Product Information
New Product
Order Now
Specificaties
Environmentally Preferable