Integral ferrule groove tools, Galtek®
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
GT12 GT4 GT8 GT6M GT2 GT8M GT6 GT12M GT5 GT10M
ENTEGT12EA 0 EUR
ENTEGT12 ENTEGT4 ENTEGT8 ENTEGT6M ENTEGT2 ENTEGT8M ENTEGT6 ENTEGT12M ENTEGT5 ENTEGT10M
Integral ferrule groove tools, Galtek®
Gereedschapset
1. Slide the ferrule nut onto the tube past the groove.
2. Insert the grooved tube end into the integral ferrule until it bottoms out.
3. Slide the ferrule nut to the fitting and hand tighten.
4. Pull on the tubing slowly until the ferrule catches on the grooved edge.
5. Ensure the fitting is hand tight.
6. Tighten the ferrule nut with a wrench. Do not overtighten.