Transport swabs
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OXOITS0001AEA 299 EUR
OXOITS0001A OXOITS0012A OXOITS0002A
Transport swabs
Wattenstaafjes Wattenstaafjes voor transport
Effective transportation of throat, vaginal, wound and skin swabs conforming to highest medical device classification - Class II a surgically invasive transient use. All swabs have soft rayon tips, which are inert and non-toxic to micro-organisms.
The addition of charcoal in the medium is considered to neutralise bacterial toxins and other inhibitory substances and has been shown to increase the recovery of Neisseria gonorrhoeae.
  • Prevents disintegration of gel column during transport
  • Eliminates undesirable air pockets, bubbles and breaks in gel which may be harmful to fastidious bacteria
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904