Manually operated diaphragm valves, Galtek® SG series
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SG4-3Q-4F SG4-3Q-4N
ENTESG4-3Q-4FEA 0 EUR
ENTESG4-3Q-4F ENTESG4-3Q-4N
Manually operated diaphragm valves, Galtek® SG series
Ventielen Microventielen
This 3-way valve (patent-pending) has an all-PFA valve body and high purity fluoropolymer wetted components providing a noncontaminating fluid stream and chemically compatible construction. Compliant to SEMI Standard F57-0301, the specification for polymer components used in ultrapure water and liquid chemical distribution systems.
  • Significantly reduces footprint, saving valuable equipment space
  • Optional integral safety lock out can lock the valve in the open or closed position
  • Stand-alone mounting base enables you to easily install and remove the valve