Combined pH electrode, HA405-DXK
Leverancier: Mettler - Toledo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
59903148 59903116 59903146
662-2124EA 0 EUR
662-2124 METN59903116 METN59903146
Combined pH electrode, HA405-DXK
Elektroden pH-elektroden
HA405-DXK is a pressure-resistant, low-maintenance pH-electrode for a variety of tough chemical applications.
  • Solid polymer electrolyte (no refilling and pressurising of electrolyte) in direct contact with the process media via an open junction
  • No clogging in contaminated media due to the open junction
  • Long operational electrode lifetime
  • Electrode allows reliable process control in highly demanding chemical processes, particulary suitable for protein-containing solutions, emulsions and suspensions, hot caustic solutions, sulfide-bearing media etc.
  • High pressure resistance: Electrode covers a generous pressure range and shows high resistance to process pressure fluctuations