Spray gun
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
221-60 215-128 215-138 221-69
ENTE221-60EA 0 EUR
ENTE221-60 ENTE215-128 ENTE215-138 ENTE221-69
Spray gun
Spuitpistolen
High purity PFA (all wetted parts).
  • With 4,8 mm orifice and ³/₈" Flaretek® tube fitting or ¹/₂" compression connections
  • Max. pressure 5,52 bar
Used in fume hoods and wet processing stations to help protect against corrosion.