Cartridge filters, Fluoroline™ S
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PFFV01D4S PFFG01D8S PFFV01D8S PFFG01D4S PFFZ01D4S PFFZ01D8S
ENTEPFFV01D4SEA 0 EUR
ENTEPFFV01D4S ENTEPFFG01D8S ENTEPFFV01D8S ENTEPFFG01D4S ENTEPFFZ01D4S ENTEPFFZ01D8S
Cartridge filters, Fluoroline™ S
Filters Filterpatronen
These cartridge filters provide small particle retention to ensure minimal particles remain on the water surface. High purity connections with no O-ring to avoid potential source of contamination ensuring downstream cleanliness.
  • Simplified installation, allows convenient retrofit with existing system and easy filter changeout
  • Manufactured in ISO 9001 quality system standard facility
Membrane: Hydrophobic PTFE; Support: PFA.
Maximum operating temperature: 60 °C
Maximum input pressure: 3,45 bar, 5 psi at 20 °C
Maximum forward/reverse differential pressure: 3,45 bar, 50 psid at 20 °C