Cartridge filters, Microgard™ UPE and UPX
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CWUG01PLT CWUG01P01 CWUG0SP01 CWUV01PLT CWUG51PLE CWUG51PLT CWUV01P01 CWUV01P01K CWUV01PLE CWUV02PLE CWUV02PLT CWUV0SP01 CWUZ01P01 CWUZ01P01K CWUZ01PLE CWUZ01PLT CWUZ01PLTK CWUZ02PLT CWUZ02PLTK CWUZ03PLT CWUZ0SP01 CWUZ51PLT CWUG01P01K CWUG01PLE CWUG02PLT
ENTECWUG01PLTEA 0 EUR
ENTECWUG01PLT ENTECWUG01P01 ENTECWUG0SP01 ENTECWUV01PLT ENTECWUG51PLE ENTECWUG51PLT ENTECWUV01P01 ENTECWUV01P01K ENTECWUV01PLE ENTECWUV02PLE ENTECWUV02PLT ENTECWUV0SP01 ENTECWUZ01P01 ENTECWUZ01P01K ENTECWUZ01PLE ENTECWUZ01PLT ENTECWUZ01PLTK ENTECWUZ02PLT ENTECWUZ02PLTK ENTECWUZ03PLT ENTECWUZ0SP01 ENTECWUZ51PLT ENTECWUG01P01K ENTECWUG01PLE ENTECWUG02PLT
Cartridge filters, Microgard™ UPE and UPX
Filters Filterpatronen
Microgard UPE/UPX filters do not require prewetting with solvent-based photochemicals. All PE construction lowers metallic and ionic contamination that can leach from filters that are manufactured with materials such as PP and nylon.
  • Particle retention rating: >99999% at stated retention rating
Membrane: Patented surface-modified hydrophilic ultra-high molecular weight polyethylene (UPE); Supports, core, sleeve: HDPE.
Maximum operating temperature: 60 °C
Maximum forward/reverse differential pressure: 3,5 bar, 50 psid at 25 °C
Maximum reverse differential pressure: 2,7 bar, 40 psid at 25 °C