Pneumatically operated diaphragm valves, Integra®
Leverancier: Entegris
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
INTEGRA™
ENTE202-86-01EA 0 EUR
ENTE202-86-01
Pneumatically operated diaphragm valves, Integra®
Ventielen Microventielen
Available compliant to SEMI Standard F57-0312, the specification for polymer components used in ultrapure water and liquid chemical distribution systems. Three-ported directional valve enables simplification of piping system design while minimising its footprint. All PFA wetted surfaces for ultrapure and corrosion resistant performance.
  • No external metal parts.
  • Unique top loading design provides for easy field service
  • Valve body design allows significant cavity drainage to minimize fluid entrapment
  • Integrally molded Flaretek® flared tube and PureBond® fusible pipe port connection options
  • Adding Espy® sensors allows for electronic monitoring of valve position
¹/₄" orifice, 3-way, position indication option.