Quantus®, Underwear Sweatshirts
Leverancier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
113-9176EA 0 EUR
113-9176 113-9169 113-9170 113-9179 113-9173 113-9177
Quantus®, Underwear Sweatshirts
Werkkleding
The Quantus® underwear sweatshirt is made from a high-quality breathable polyester microfibre material with carbon. The fabric was specially developed for use as underwear in combination with a cleanroom overall or coat. It serves as a prefilter to allow open-pore-fabric to be used in a higher cleanroom class.
  • Sweatshirt with long sleeves and round neck
  • Extra material flap for bar code inside
  • Soft grip and greater comfort when wearing
  • Anti-static and antimicrobial protection
  • Recommended for wear underneath disposable and reusable clothing
  • RFID chip included
  • Washing instructions: max 60 °C
Weight: 97,5 g/m²
Bestelinformatie: Non standard sizes are available on request. Please contact your local Avantor office for further information.