Mask packages, H 60 series
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
H60-50-0615 H60-3035-0615 H60-25-0615
ENTEH60-50-0615EA 0 EUR
ENTEH60-50-0615 ENTEH60-3035-0615 ENTEH60-25-0615
Mask packages, H 60 series
Wafertransportsystemen Wafer dragers
Holds one mask or reticle.
  • Manufactured and packaged in a cleanroom environment
  • Bagged and sealed in a cleanroom bag and heat sealed in a second polyethylene bag
  • Living hinge
  • Compact and stackable
  • Dust-proof