Wafer storage boxes for 76,2 mm (3") wafer handling, E 52-101 series
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E52-101-61C02 E96-2-1613 E96-2P-21A15
ENTEE52-101-61C02EA 0 EUR
ENTEE52-101-61C02 ENTEE96-2-1613 ENTEE96-2P-21A15
Wafer storage boxes for 76,2 mm (3") wafer handling, E 52-101 series
Wafertransportsystemen Wafer Opslagdozen
  • Angeled box bottom
  • Pin and socket hinges
  • Three swing pin latches
  • Stackable
  • Two carrier capacity
  • Compatible carriers: all 3" wafer carriers
  • Overall size: 203×236×114 mm