Wafer storage boxes for 100 mm (4") wafer handling, E 97 series
Leverancier: ENTEGRIS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
E97-102-61C02 E97-102-0603 E97-101-0603 E97-101-0604 E97-101-0605 E97-101-0615
ENTEE97-102-61C02EA 0 EUR
ENTEE97-102-61C02 ENTEE97-102-0603 ENTEE97-101-0603 ENTEE97-101-0604 ENTEE97-101-0605 ENTEE97-101-0615
Wafer storage boxes for 100 mm (4") wafer handling, E 97 series
Wafertransportsystemen Wafer Opslagdozen
With pin and socket hinges. Single snap latch.
  • Stackable
  • Optional wafer cushion
  • Single carrier capacity
  • Compatible carriers: 182 series, 'M' style 72 series