picoLUCENT™ PLUS-AP kit
Leverancier: G Bioscienes
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
786-09AP 786-09APT
786-09APEA 541 EUR
786-09AP 786-09APT
picoLUCENT™ PLUS-AP kit
Reagentia voor elektroforese Western Blotting Reagents
The picoLUCENT™ PLUS-HRP kit is based on an ultra sensitive luminol substrate that produces chemiluminescence upon reaction with alkaline phosphatase. The chemiluminescence light emission can be recorded by a short exposure to autoradiography films.
  • Low picogram detection (10⁻¹²) of antigens
  • Low noise (signal/background ratio)
Leveringsinformatie: Primary antibody not included.