Powder spatulas, disposable, SteriWare® PowderSafe
Leverancier: Sampling Systems
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
8097A-150S 8097A-150
231-0444EA 495 EUR
231-0444 231-0443
Powder spatulas, disposable, SteriWare® PowderSafe
Spatels
PS, white. These disposable powder spatulas come complete with a click-on container.
  • Sampled powder is safely locked inside the cover
  • Cross-contamination risk is eliminated - the device can be used straight from the bag
  • Manufactured in a medical cleanroom environment
  • Approved for use in ATEX Zones 0, 1 and 2
The transparent container slips over the blade of the spatula and is securely locked into place. At the laboratory the container can be easily removed. Suitable for all industries (pharmaceutical, food, chemical, cosmetic, biotechnology etc.)