Shelving starter and add-on units, Super Erecta®
Leverancier: Metro International
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
5A347C 5A357C 5N317C
METO5A347CEA 856 EUR
METO5A347C METO5A357C METO5N317C
Shelving starter and add-on units, Super Erecta®
Meubilair Legborden
These Super Erecta® Starter save time and money by eliminating adjacent posts and replacing them with 'S' hooks. This system offers a convenient way to create long runs of shelving, shelving at right angles and combinations of runs and right angles.
The load capacity evenly distributed for shelf lengths up to 1219 mm is 363 kg per shelf and for shelf lengths 1524 mm and longer is 272 kg per shelf.
Chrome plated starter units includes four 1880 mm post and five 457 mm deep Super Erecta® shelves.