Peqlab peqGOLD, DNA Ladder, 100 bp
Leverancier: VWR
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Peqlab peqGOLD
25-2010EA 73.6 EUR
25-2010
Peqlab peqGOLD, DNA Ladder, 100 bp
Reagentia voor elektroforese Molecuulgewichtmerkers
DNA ladder for quantitative and qualitative analysis of dsDNA.
  • Range: 10 fragments, 100 to 1000 bp in size
  • Highlighted 500 bp band for easy orientation
  • Also available as ready to use mix containing Orange G dye instead of bromophenol blue