Oils for rotary vane pumps
Leverancier: Gardner Denver
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
ILMVU8995G-11EA 83.1 EUR
ILMVU8995G-11 ILMV8995G-15 ILMV8995G-11 ILMV8995G-20
Oils for rotary vane pumps
Smeermiddelen
This double distilled synthetic hydrocarbon oil designed for excellent resistance when pumping corrosive gases or vapors. A synthetic base stock that has no aromatic compounds or sulfur which accelerate varnishing, sludging and carbon build-up when pumps are used to pump corrosives.
  • Exceptionally low vapor pressure
  • High stability in chemical environments
  • No additives or inhibitors