Glass microfiber filters without binder, grade MN 85/90 BF
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
406024 4060037 406015 4060025 4060045 406005 406007 406009 406011 406012 406018 406027 406014
MANA406024EA 377 EUR
MANA406024 MANA4060037 MANA406015 MANA4060025 MANA4060045 MANA406005 MANA406007 MANA406009 MANA406011 MANA406012 MANA406018 MANA406027 MANA406014
Glass microfiber filters without binder, grade MN 85/90 BF
Filters Glasvezelfilters
This paper is especially used in water treatment facilities for example for the determination of suspended solids.
  • For difference weight analysis
  • Binder-free
  • Suitable for suspended solids analysis
Order Now
Specificaties
Environmentally Preferable
Gewicht 90 g/m²
Dikte 0,40 mm
Partikelretentie 0,5 μm
Filtratiesnelheid 15 s (air)
Maximumtemperatuur 500 °C