Mesh woven filters, nylon, Spectra Mesh®
Leverancier: Spectrum Laboratories
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
146498 146506 146514 145811 146490 146488 148146 145914 146479 145813 148134 148130 148131 148132 145925 145908 145807 146487 146494 148140 146502 145922 145920 145916 146486 148143 148145 146510 146519 148138 148136 145801 145803 145816
SPEC146498EA 403 EUR
SPEC146498 SPEC146506 SPEC146514 SPEC145811 SPEC146490 SPEC146488 SPEC148146 SPEC145914 SPEC146479 SPEC145813 SPEC148134 SPEC148130 SPEC148131 SPEC148132 SPEC145925 SPEC145908 SPEC145807 SPEC146487 SPEC146494 SPEC148140 SPEC146502 SPEC145922 SPEC145920 SPEC145916 SPEC146486 SPEC148143 SPEC148145 SPEC146510 SPEC146519 SPEC148138 SPEC148136 SPEC145801 SPEC145803 SPEC145816
Mesh woven filters, nylon, Spectra Mesh®
Filters Membraanfilters
These woven filters are constructed of individual strands woven into a mesh screen, characterised by precise mesh openings, percent open area and mesh thickness. Available in five material types with different physical, chemical and thermal characteristics to select from depending on your application. Available in discs or square sheets.
Water affinity: Hydrophilic
Strength/durability: High
Adsorption: High
Best resistance: Corrosives
pH resistance: 3 to 10
Thermal stability: up to 180 °C
Sterilisation: Irradiation