Mesh woven filters, polyester, Spectra Mesh®
Leverancier: Spectrum Laboratories
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
146521 148282 148283 145840 146524 146525 146526 146530
734-3518EA 375 EUR
734-3518 734-3560 734-3561 734-3529 734-3542 734-3543 734-3544 734-3545
Mesh woven filters, polyester, Spectra Mesh®
Filters Membraanfilters
These woven filters are constructed of individual strands woven into a mesh screen, characterised by precise mesh openings, percent open area and mesh thickness. Available in five material types with different physical, chemical and thermal characteristics to select from depending on your application. Available in discs or square sheets.
Water affinity: Mildly hydrophobic
Strength/durability: High
Adsorption: Low
Best resistance: Alkalis / organics
pH resistance: 3 to 13
Thermal stability: up to 140 ºC
Sterilisation: Autoclavable