Neonate heads
Leverancier: 3B Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
1005102 1005531
BINH1005102EA 0 EUR
BINH1005102 BINH1005531
Neonate heads
Modellen Menselijke biologie modellen
This neonatal intubation trainer provides for practice of intubation skills on a newborn baby.
  • Realistic anatomy
  • Oral and nasal intubation, bag valve mask ventilation
  • Correct tube placement can be checked by practical inflation tests
Robust and realistic, this model allows students to undertake training that is directly transferable to the clinical setting.