Peqlab peqGREEN, DNA/RNA Dye
Leverancier: Peqlab brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
732-3196EA 185 EUR
732-3196 732-2960
Peqlab peqGREEN, DNA/RNA Dye
Kleurstoffen Kleuringen voor elektroforese
peqGREEN dye is a safe alternative to ethidium bromide for staining of dsDNA, ssDNA and RNA.
  • Direct addition to gel solution or staining after electrophoresis
  • Excitation with UV or blue light
Suitable for up to 400 gels.