Control Cables
Leverancier: HUBER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
6146 6279 9353 55018 9490 9491 9492 54949
461-0622EA 155 EUR
461-0622 HUBE6279 HUBE9353 HUBE55018 HUBE9490 HUBE9491 HUBE9492 HUBE54949
Control Cables
Kabels
A range of control cables is available for USB, RS232, RS485 or analogue interfaces (AIF). Cables are also available for external control signal (ECS), a potentialfree contact (POKO) or for an external float switch (LEVEL).